Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-Carnitine hydrochloride, ≥97.0% (HPLC), MilliporeSigma™ Supelco™
$153.00
Specifications
CAS | 6645-46-1 |
---|---|
Type | Analytical Standard |
Linear Formula | (CH3)3 N+CH2 CH(OH)CH2 CO2 H · Cl- |
MDL Number | MFCD00066100 |
Synonym | (-)-beta-Hydroxy-gamma-(trimethylammonio)butyrate; Vitamin BT |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
11-101-5116
|
MilliporeSigma Supelco
9495450MG |
50 mg |
Each of 1 for $153.00
|
|
|||||
Specifications
6645-46-1 | |
(CH3)3 N+CH2 CH(OH)CH2 CO2 H · Cl- | |
(-)-beta-Hydroxy-gamma-(trimethylammonio)butyrate; Vitamin BT | |
≥97.0% (HPLC) |
Analytical Standard | |
MFCD00066100 | |
197.66 | |
Analytical Standard |
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title