Learn More
Tocris Bioscience™ L-CCG-lll
Potent, competitive glutamate uptake inhibitor
Manufacturer:  Tocris Bioscience™ 0332/10
 View more versions of this product
Catalog No. 03-321-0
  Check Availability
Add to cart
Description
Description
Potent and competitive inhibitor of both glial and neuronal uptake of glutamate, aspartate and cysteate.
Specifications
Specifications
(2S,1'S,2'R)-2-(Carboxycyclopropyl)glycine
C6H9NO4
10mg
117857-95-1
Observed MW: 159.14
Soluble to 100 mM in 1eq. NaOH
Safety and Handling
Safety and Handling
Recommended Storage : Store at RT