Learn More
Tocris Bioscience™ L-CCG-lll Available on GSA/VA Contract for Federal Government customers only.
Potent, competitive glutamate uptake inhibitor
Manufacturer:  Tocris Bioscience™ 0332/5
 View more versions of this product
Catalog No. 03-325
  Check Availability
Add to cart
Description
Description
Potent and competitive inhibitor of both glial and neuronal uptake of glutamate, aspartate and cysteate.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
(2S,1'S,2'R)-2-(Carboxycyclopropyl)glycine
C6H9NO4
5mg
117857-95-1
Observed MW: 159.14
Soluble to 100 mM in 1eq. NaOH
Safety and Handling
Safety and Handling
Recommended Storage : Store at RT