Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Cyclopentylglycine, 95%

Click to view available options
Quantity:
250mg
1g
5g
2521-84-8
C7H13NO2
143.186
MFCD03788076
XBPKRVHTESHFAA-LURJTMIESA-N
l-cyclopentylglycine,s-2-amino-2-cyclopentylacetic acid,2-cyclopentyl-l-glycine,h-cyclopentyl-gly-oh,l-beta-cyclopentylglycine,2s-amino cyclopentyl ethanoic acid,h-l-cpg-oh,s-2-cyclopentylglycine,s-amino-cyclopentyl-acetic acid,2s-2-amino-2-cyclopentylacetic acid
10374564
(2S)-2-amino-2-cyclopentylacetic acid
C1CCC(C1)C(C(=O)O)N
2521-84-8
C7H13NO2
143.186
MFCD03788076
XBPKRVHTESHFAA-LURJTMIESA-N
l-cyclopentylglycine,s-2-amino-2-cyclopentylacetic acid,2-cyclopentyl-l-glycine,h-cyclopentyl-gly-oh,l-beta-cyclopentylglycine,2s-amino cyclopentyl ethanoic acid,h-l-cpg-oh,s-2-cyclopentylglycine,s-amino-cyclopentyl-acetic acid,2s-2-amino-2-cyclopentylacetic acid
10374564
(2S)-2-amino-2-cyclopentylacetic acid
C1CCC(C1)C(C(=O)O)N
Formula Weight 143.18
Percent Purity 95%
Quantity 250mg
Chemical Name or Material L-Cyclopentylglycine

TSCA : No

Recommended Storage : Ambient temperatures