Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ L-Cysteine Hydrochloride Monohydrate, 98.5 to 101.5% (Dry Basis), Thermo Scientific™

Click to view available options
Quantity:
25g
100g
1kg
5kg
Packaging:
Plastic bottle
7048-04-6
C3H10ClNO3S
175.63
MFCD00065606
QIJRTFXNRTXDIP-JIZZDEOASA-N
l-cysteine hydrochloride monohydrate,h-cys-oh.hcl.h2o,l-cysteine hydrochloride hydrate,l-cysteine monohydrate monochloride,r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate,unii-zt934n0x4w,l-cysteine hydrate hydrochloride,cysteine hydrochloride monohydrate, l,h-cys-ohhclh2o,cysteine hcl
23462
(2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
C(C(C(=O)O)N)S.O.Cl
7048-04-6
C3H10ClNO3S
175.63
MFCD00065606
QIJRTFXNRTXDIP-JIZZDEOASA-N
l-cysteine hydrochloride monohydrate,h-cys-oh.hcl.h2o,l-cysteine hydrochloride hydrate,l-cysteine monohydrate monochloride,r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate,unii-zt934n0x4w,l-cysteine hydrate hydrochloride,cysteine hydrochloride monohydrate, l,h-cys-ohhclh2o,cysteine hcl
23462
(2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
C(C(C(=O)O)N)S.O.Cl
Linear Formula HSCH2CH(NH2)COOH.HCl.H2O
Packaging Plastic bottle
Formula Weight 175.63
Physical Form Crystalline Powder
Percent Purity 98.5 to 101.5%
Melting Point ~175°C (decomposition)
Quantity 5kg
Chemical Name or Material L-Cysteine Hydrochloride Monohydrate
Assay Dry basis