Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Cysteine Hydrochloride, Monohydrate, Multi-Compendial, U.S.P., J.T.Baker™

Click to view available options
Quantity:
100g
1kg
Packaging:
Poly Bottle

Chemical Identifiers

7048-04-6
C3H10ClNO3S
175.63
QIJRTFXNRTXDIP-JIZZDEOASA-N
23462
CAS 7048-04-6
Molecular Formula C3H10ClNO3S
Packaging Poly Bottle
InChI Key QIJRTFXNRTXDIP-JIZZDEOASA-N
SMILES C(C(C(=O)O)N)S.O.Cl
IUPAC Name (2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
Molecular Weight (g/mol) 175.63
PubChem CID 23462
Formula Weight 175.64
Quantity 100g
Specific Gravity 1L = 1.50kg
Chemical Name or Material L-Cysteine Hydrochloride, Monohydrate
Grade Multi-Compendial/USP
Show More Show Less