Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Cystine, 98.5 to 101.5% (Dry Basis), Affymetrix/USB™

Click to view available options
Quantity:
50g
1kg
5kg
Packaging:
Plastic bottle
56-89-3
C6H12N2O4S2
240.292
MFCD00064228
LEVWYRKDKASIDU-IMJSIDKUSA-N
l-cystine,cystine,l-dicysteine,l-cystin,cystine acid,cysteine disulfide,oxidized l-cysteine,beta,beta'-dithiodialanine,h-cys-oh 2,l-cysteine disulfide
67678
CHEBI:16283
(2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid
C(C(C(=O)O)N)SSCC(C(=O)O)N
56-89-3
C6H12N2O4S2
240.292
MFCD00064228
LEVWYRKDKASIDU-IMJSIDKUSA-N
l-cystine,cystine,l-dicysteine,l-cystin,cystine acid,cysteine disulfide,oxidized l-cysteine,beta,beta'-dithiodialanine,h-cys-oh 2,l-cysteine disulfide
67678
CHEBI:16283
(2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid
C(C(C(=O)O)N)SSCC(C(=O)O)N
Packaging Plastic bottle
Formula Weight 240.30
Physical Form Crystalline Powder
Percent Purity 98.5 to 101.5%
Melting Point >240°C (decomposition)
Quantity 5kg
Specific Gravity 1.68
Chemical Name or Material L-Cystine