Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-Cystine dihydrochloride, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
250 g
Description
Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.
Solubility
Soluble in hydrochloric acid. (0.5 M HCl: 50 mg/mL) Soluble in water.
Notes
Store at room temperature. Incompatible with oxidizing agents.
Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.
Solubility
Soluble in hydrochloric acid. (0.5 M HCl: 50 mg/mL) Soluble in water.
Notes
Store at room temperature. Incompatible with oxidizing agents.
Specifications
Melting Point | 228°C to 232°C |
Color | White to Yellow |
Quantity | 100 g |
Solubility Information | Soluble in hydrochloric acid. (0.5 M HCl: 50mg/mL) Soluble in water. |
Formula Weight | 313.22 |
Percent Purity | 99% |
Physical Form | Powder |
Chemical Name or Material | L-Cystine dihydrochloride |
Safety and Handling
EINECSNumber | 250-391-9 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction