Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-(-)-Cystine 98.0+%, TCI America™

Click to view available options
Quantity:
25 g
500 g
56-89-3
C6H12N2O4S2
240.292
MFCD00064228
LEVWYRKDKASIDU-IMJSIDKUSA-N
l-cystine, cystine, l-dicysteine, l-cystin, cystine acid, cysteine disulfide, oxidized l-cysteine, beta,beta'-dithiodialanine, h-cys-oh 2, l-cysteine disulfide
67678
CHEBI:16283
(2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid
C(C(C(=O)O)N)SSCC(C(=O)O)N
56-89-3
C6H12N2O4S2
240.292
MFCD00064228
LEVWYRKDKASIDU-IMJSIDKUSA-N
l-cystine, cystine, l-dicysteine, l-cystin, cystine acid, cysteine disulfide, oxidized l-cysteine, beta,beta'-dithiodialanine, h-cys-oh 2, l-cysteine disulfide
67678
CHEBI:16283
(2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid
C(C(C(=O)O)N)SSCC(C(=O)O)N
Formula Weight 240.29
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,N)
Color White
Melting Point 261°C
Quantity 500 g
Chemical Name or Material L-(-)-Cystine

EINECSNumber : (9)-1587&(2)-1468

RTECSNumber : HA2690000

TSCA : Yes