Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Dihydroorotic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
5988-19-2
C5H6N2O4
158.11
UFIVEPVSAGBUSI-REOHCLBHSA-N
l-dihydroorotic acid,s-2,6-dioxohexahydropyrimidine-4-carboxylic acid,l-dihydroorotate,dihydro-l-orotic acid,s-dihydroorotic acid,l-hydroorotic acid,s-dihydroorotate,hydroorotic acid, l,4s-2,6-dioxo-1,3-diazinane-4-carboxylic acid,s-4,5-dihydroorotate
439216
CHEBI:17025
(4S)-2,6-dioxo-1,3-diazinane-4-carboxylic acid
C1C(NC(=O)NC1=O)C(=O)O
5988-19-2
C5H6N2O4
158.11
UFIVEPVSAGBUSI-REOHCLBHSA-N
l-dihydroorotic acid,s-2,6-dioxohexahydropyrimidine-4-carboxylic acid,l-dihydroorotate,dihydro-l-orotic acid,s-dihydroorotic acid,l-hydroorotic acid,s-dihydroorotate,hydroorotic acid, l,4s-2,6-dioxo-1,3-diazinane-4-carboxylic acid,s-4,5-dihydroorotate
439216
CHEBI:17025
(4S)-2,6-dioxo-1,3-diazinane-4-carboxylic acid
C1C(NC(=O)NC1=O)C(=O)O
Infrared Spectrum Authentic
Melting Point 254.0°C to 255.0°C
Packaging Glass bottle
Quantity 5g
Color White
Assay Percent Range 98.5% min. (HPLC)
Formula Weight 158.11
Physical Form Powder
Percent Purity 99%
Chemical Name or Material L-Dihydroorotic acid

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

GHS Signal Word: Warning