Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L(-)-Epinephrine, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
51-43-4
C9H13NO3
183.21
MFCD00002204
UCTWMZQNUQWSLP-VIFPVBQESA-N
epinephrine,l-adrenaline,adrenaline,l-epinephrine,adrenalin,levoepinephrine,epinefrina,--adrenaline,adnephrine,chelafrin
5816
CHEBI:28918
4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
CNCC(C1=CC(=C(C=C1)O)O)O
51-43-4
C9H13NO3
183.21
MFCD00002204
UCTWMZQNUQWSLP-VIFPVBQESA-N
epinephrine,l-adrenaline,adrenaline,l-epinephrine,adrenalin,levoepinephrine,epinefrina,--adrenaline,adnephrine,chelafrin
5816
CHEBI:28918
4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
CNCC(C1=CC(=C(C=C1)O)O)O
Specific Rotation Condition − 51.50 (20.00°C c=4,1M HCl,dry sub)
Packaging Glass bottle
Color White to Beige
Sulfated Ash 0.1% max.
Specific Rotation − 51.50
Infrared Spectrum Authentic
Quantity 25g
Loss on Drying 1% max. (18 hrs) (0.7 kPa, P2O5)
Melting Point 208.0°C to 211.0°C
Assay Percent Range 99%
Linear Formula (HO)2C6H3CH(CH2NHCH3)OH
Beilstein 13, 830
Merck Index 15, 3674
Solubility Information Solubility in water: sparingly soluble. Other solubilities: insoluble in alcohol, chloroform, ether and, acetone
Formula Weight 183.21
Physical Form Fine Crystalline Powder
Percent Purity 99%
Chemical Name or Material L(-)-Epinephrine, 99%
Show More Show Less

GHS H Statement
Causes skin irritation. Toxic if inhaled. Toxic if swallowed. Toxic in contact with skin. Causes serious eye irritation. May cause respiratory irritation. Harmful to aquatic life with long lasting effects. May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS Signal Word: Danger

EINECSNumber : 200-098-7

RTECSNumber : DO2625000

TSCA : TSCA

Recommended Storage : product darkens on exposure to air and light