Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L(-)-Fenchone, 98+%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
Packaging:
Glass bottle
7787-20-4
C10H16O
152.24
MFCD00151104
LHXDLQBQYFFVNW-GMSGAONNSA-N
l-fenchone,1r-fenchone,1r,4r-2,2,4-trimethylbicyclo 2.2.1 heptan-3-one,2r,5r-2,6,6-trimethyl-2,5-methanocyclohexane-1-one
3034206
(1R,4R)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one
CC1(C2CCC(C2)(C1=O)C)C
7787-20-4
C10H16O
152.24
MFCD00151104
LHXDLQBQYFFVNW-GMSGAONNSA-N
l-fenchone,1r-fenchone,1r,4r-2,2,4-trimethylbicyclo 2.2.1 heptan-3-one,2r,5r-2,6,6-trimethyl-2,5-methanocyclohexane-1-one
3034206
(1R,4R)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one
CC1(C2CCC(C2)(C1=O)C)C
Boiling Point 192°C to 194°C
Melting Point 5°C
Flash Point 52°C
Specific Gravity 0.94
Density 0.9400g/mL
Specific Rotation -50.5
Refractive Index 1.4610 to 1.463
Color Colorless to Yellow
Infrared Spectrum Authentic
Specific Rotation Condition −50.50° (24°C neat)
Packaging Glass bottle
Quantity 50g
Assay Percent Range 98% min. (GC)
Fieser 08,228
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether
Formula Weight 152.24
Physical Form Liquid
Percent Purity 98+%
Chemical Name or Material L(-)-Fenchone, 98+%
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 232-107-5

TSCA : TSCA