Learn More
Invitrogen™ L-Ficolin Monoclonal Antibody (19)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ ABS0051902
Description
ABS 005-19-02 detects L-Ficolin from human samples. This antibody is specific towards L-Ficolin; no cross reactivity is seen with M-Ficolin (Ficolin-1) and H-Ficolin (Ficolin-3). ABS 005-19-02 has been successfully used in ELISA and WB procedures. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.
L-Ficolin (Ficolin-2) is an innate immunity pattern recognition molecule found in human serum. L-Ficolin binds to distinct pathogen-associated molecular patterns (PAMP), such as carbohydrates (GlcNAc), lipoteichoic acid and acetylated groups thereby facilitating phagocytosis and activation of complement through the lectin route using the same serine proteases as mannose-binding lectin (MBL) named MASP ((MBL)-associated serine protease).
Specifications
L-Ficolin | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
Q15485 | |
FCN2 | |
Purified recombinant protein corresponding to human L-Ficolin and full-length L-Ficolin (Ficolin-2). | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunoprecipitation, Western Blot | |
19 | |
Unconjugated | |
FCN2 | |
37 kDa elastin-binding protein; Collagen/fibrinogen domain-containing protein 2; EBP-37; FCN2; Fcnb; FCNL; ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin); ficolin 2; ficolin B; ficolin-2; Ficolin-B; ficolin-beta; Hucolin; L-ficolin; P35; RP11-263F14.2; Serum lectin p35 | |
Mouse | |
Protein A/G | |
RUO | |
2220 | |
4°C, store in dark | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.