Learn More
Chem-Impex International, Inc.Supplier Diversity Partner L(-)-Fucose, 98 - 100% (HPLC), CAS#: 6696-41-9 / 2438-80-4, 25G DFS Item
Manufacturer:  Chem-Impex International, Inc.Supplier Diversity Partner 0073625G
L(-)-Fucose, 98 - 100% (HPLC), CAS#: 6696-41-9 / 2438-80-4, 25G. Hazardous Material: NO. MDL #: MFCD00069812. Molecular Formula: C6H12O5. Molecular Weight: 164.16. Melting Point: 150 - 153 °C. Appearance: White to off-white crystalline powder. Optical Rotation: [a]20D = -75 ± 2 º (C=4 in H2O). Storage Temperature: Store at RT.
Catalog No. 50-492-171