Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Glutamic acid 1-tert-butyl ester, 97%

Click to view available options
Quantity:
250mg
1g
5g
45120-30-7
C9H17NO4
203.238
MFCD00038562
QVAQMUAKTNUNLN-LURJTMIESA-N
h-glu-otbu,s-4-amino-5-tert-butoxy-5-oxopentanoic acid,1-tert-butyl l-glutamate,l-glutamic acid 1-tert-butyl ester,4s-4-amino-5-tert-butoxy-5-oxopentanoic acid,glu-otbu,h-gluotbu,ambotzhaa5210,pubchem17225,glutamic acid t-butyl ester
117505
(4S)-4-amino-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
CC(C)(C)OC(=O)C(CCC(=O)O)N
45120-30-7
C9H17NO4
203.238
MFCD00038562
QVAQMUAKTNUNLN-LURJTMIESA-N
h-glu-otbu,s-4-amino-5-tert-butoxy-5-oxopentanoic acid,1-tert-butyl l-glutamate,l-glutamic acid 1-tert-butyl ester,4s-4-amino-5-tert-butoxy-5-oxopentanoic acid,glu-otbu,h-gluotbu,ambotzhaa5210,pubchem17225,glutamic acid t-butyl ester
117505
(4S)-4-amino-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
CC(C)(C)OC(=O)C(CCC(=O)O)N
Beilstein 2414793
Formula Weight 203.24
Physical Form Powder
Percent Purity 97%
Color White
Melting Point 139°C to 145°C
Quantity 250mg
Chemical Name or Material L-Glutamic acid 1-tert-butyl ester

storageNote1 : 2° to 8°C