Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ L-Glutamic acid, 98.5 to 101.5% (Dry Basis), Ultrapure, Thermo Scientific™

Click to view available options
Quantity:
1kg
5kg
25kg
Packaging:
Plastic bottle
56-86-0
C5H9NO4
147.13
MFCD00002634
WHUUTDBJXJRKMK-VKHMYHEASA-N
l-glutamic acid,glutamic acid,2s-2-aminopentanedioic acid,h-glu-oh,s-2-aminopentanedioic acid,glutaminol,l-glutaminic acid,glutacid,glutamicol,glutamidex
33032
CHEBI:16015
(2S)-2-aminopentanedioic acid
C(CC(=O)O)C(C(=O)O)N
56-86-0
C5H9NO4
147.13
MFCD00002634
WHUUTDBJXJRKMK-VKHMYHEASA-N
l-glutamic acid,glutamic acid,2s-2-aminopentanedioic acid,h-glu-oh,s-2-aminopentanedioic acid,glutaminol,l-glutaminic acid,glutacid,glutamicol,glutamidex
33032
CHEBI:16015
(2S)-2-aminopentanedioic acid
C(CC(=O)O)C(C(=O)O)N
Linear Formula HOOCCH2CH2CH(NH2)COOH
Packaging Plastic bottle
Formula Weight 147.13
Physical Form Crystalline Powder
Percent Purity 98.5 to 101.5%
Melting Point 189°C to 192°C (decomposition)
Quantity 5kg
Specific Gravity 1.525
Chemical Name or Material L-Glutamic acid
Assay Dry basis