Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-(+)-Glutamic Acid, BAKER ANALYZED™ Biochemical Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
1kg
Packaging:
Poly Bottle

Chemical Identifiers

56-86-0
C5H9NO4
147.13
WHUUTDBJXJRKMK-VKHMYHEASA-N
33032
CHEBI:16015
(2S)-2-aminopentanedioic acid
C(CC(=O)O)C(C(=O)O)N

High quality biochemicals for research use

Packaging Poly Bottle
Formula Weight 147.13
Quantity 1kg
Specific Gravity 1L = 1.54kg
Chemical Name or Material L-(+)-Glutamic Acid
Grade BAKER ANALYZED™ Biochemical Reagent