Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Glutamic acid dimethyl ester hydrochloride, 99%, pure, Acros Organics

Click to view available options
Quantity:
10g
50g
23150-65-4
C7H13NO4·ClH
211.65
MFUPLHQOVIUESQ-JEDNCBNOSA-N
l-glutamic acid dimethyl ester hydrochloride,h-glu ome-ome.hcl,dimethyl l-glutamate hydrochloride,s-dimethyl 2-aminopentanedioate hydrochloride,unii-185vc5x3pf,1,5-dimethyl 2s-2-aminopentanedioate hydrochloride,h-glu ome-ome inverted exclamation mark currencyhcl,s-dimethyl 2-aminopentanedioate hcl,glutamic acid dimethyl ester hydrochloride,dimethyl glutamic acid hydrochloride
12917568
dimethyl (2S)-2-aminopentanedioate;hydrochloride
COC(=O)CCC(C(=O)OC)N.Cl
23150-65-4
C7H13NO4·ClH
211.65
MFUPLHQOVIUESQ-JEDNCBNOSA-N
l-glutamic acid dimethyl ester hydrochloride,h-glu ome-ome.hcl,dimethyl l-glutamate hydrochloride,s-dimethyl 2-aminopentanedioate hydrochloride,unii-185vc5x3pf,1,5-dimethyl 2s-2-aminopentanedioate hydrochloride,h-glu ome-ome inverted exclamation mark currencyhcl,s-dimethyl 2-aminopentanedioate hcl,glutamic acid dimethyl ester hydrochloride,dimethyl glutamic acid hydrochloride
12917568
dimethyl (2S)-2-aminopentanedioate;hydrochloride
COC(=O)CCC(C(=O)OC)N.Cl
Specific Rotation 26
Melting Point 91°C to 93°C
Color White
Infrared Spectrum Authentic
Quantity 10g
Packaging Glass bottle
Assay Percent Range 98.5% min. (Argentometry)
Formula Weight 211.65
Physical Form Powder
Percent Purity 99%
Grade Pure
Chemical Name or Material L-Glutamic acid dimethyl ester hydrochloride
Show More Show Less

EINECSNumber : 245-461-