Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L(+)-Glutamine, Acros Organics

Click to view available options
Quantity:
50g
250g
1kg
56-85-9
C5H10N2O3
146.146
ZDXPYRJPNDTMRX-VKHMYHEASA-N
l-glutamine,glutamine,levoglutamide,l-+-glutamine,glutamic acid amide,h-gln-oh,stimulina,cebrogen,glumin,levoglutamid
5961
CHEBI:18050
(2S)-2,5-diamino-5-oxopentanoic acid
C(CC(=O)N)C(C(=O)O)N
56-85-9
C5H10N2O3
146.146
ZDXPYRJPNDTMRX-VKHMYHEASA-N
l-glutamine,glutamine,levoglutamide,l-+-glutamine,glutamic acid amide,h-gln-oh,stimulina,cebrogen,glumin,levoglutamid
5961
CHEBI:18050
(2S)-2,5-diamino-5-oxopentanoic acid
C(CC(=O)N)C(C(=O)O)N
Linear Formula H2NCOCH2CH2CH(NH2)CO2H
Formula Weight 146.15
Solubility Solubility in water: 35g/L (20°C). Other solubilities: practically insoluble in methanol, ethanol, ether,, benzene, acetone, ethyl acetate, chloroform, soluble in mineral acids, soluble in ethanol: 46 μg/100mL (25°C), soluble in methanol: 3.5 mg/100mL (2
Melting Point 185.0°C
Quantity 250g
Chemical Name or Material L(+)-Glutamine