Learn More
UbpbioSupplier Diversity Partner L-GLUTATHIONE, 25G DFS Item
Manufacturer:  UbpbioSupplier Diversity Partner P103025
Protein Purification Reagents – Reduced L-Glutathione (UBPBio catalog# P1030-25, P1030-100) – Reduced L-glutathione is a natural compound made in the body from the amino acids glutamic acid, cysteine, and glycine. As the body’s major cellular antioxidant, reduced L-glutathione is crucial for scavenging damaging free radicals. It is also used for eluting GST-tagged proteins from glutathione resins.
Catalog No. 50-112-6939