Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Histidinamide dihydrochloride, 95%

Click to view available options
Quantity:
250mg
1g
5g
71666-95-0
C6H12Cl2N4O
227.089
MFCD00058559
CRBYFJCXMZNLTO-XRIGFGBMSA-N
h-his-nh2.2hcl,l-histidinamide dihydrochloride,h-his-nh2 2hcl,h-his-nh 2 hcl,c6h10n4o.2hcl,h-his-nh2 . 2 hcl,l-histidine amide dihydrochloride,l-histidinamide-hydrogen chloride 1/2,h-his-nh2 inverted exclamation mark currency 2 hcl,2s-2-amino-3-1h-imidazol-4-yl propanamide dihydrochloride
56777274
(2S)-2-amino-3-(1H-imidazol-5-yl)propanamide;dihydrochloride
C1=C(NC=N1)CC(C(=O)N)N.Cl.Cl
71666-95-0
C6H12Cl2N4O
227.089
MFCD00058559
CRBYFJCXMZNLTO-XRIGFGBMSA-N
h-his-nh2.2hcl,l-histidinamide dihydrochloride,h-his-nh2 2hcl,h-his-nh 2 hcl,c6h10n4o.2hcl,h-his-nh2 . 2 hcl,l-histidine amide dihydrochloride,l-histidinamide-hydrogen chloride 1/2,h-his-nh2 inverted exclamation mark currency 2 hcl,2s-2-amino-3-1h-imidazol-4-yl propanamide dihydrochloride
56777274
(2S)-2-amino-3-(1H-imidazol-5-yl)propanamide;dihydrochloride
C1=C(NC=N1)CC(C(=O)N)N.Cl.Cl
Beilstein 3720223
Formula Weight 227.09
Physical Form Solid
Percent Purity 95%
Quantity 250mg
Chemical Name or Material L-Histidinamide dihydrochloride

storageNote1 : 2° to 8°C