Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Histidine Monohydrochloride Monohydrate, ≥98%, Thermo Scientific™

Catalog Number aaj1708022
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic Bottle
5934-29-2
C6H12ClN3O3
209.63
MFCD00151027
CMXXUDSWGMGYLZ-XRIGFGBMSA-N
l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1 h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.H2O, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate
165377
(2 S)-2-amino-3-(1 H-imidazol-5-yl)propanoic acid;hydrate;hydrochloride
C1=C(NC=N1)CC(C(=O)O)N.O.Cl
5934-29-2
C6H12ClN3O3
209.63
MFCD00151027
CMXXUDSWGMGYLZ-XRIGFGBMSA-N
l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1 h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.H2O, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate
165377
(2 S)-2-amino-3-(1 H-imidazol-5-yl)propanoic acid;hydrate;hydrochloride
C1=C(NC=N1)CC(C(=O)O)N.O.Cl
Melting Point ∼240°C (decomposition)
Linear Formula (C3H3N2)CH2CH(NH2)COOH.HCl.H2O
Beilstein 4168261
Packaging Plastic Bottle
Formula Weight 209.63
Percent Purity ≥98%
Quantity 100 g
Physical Form Crystalline Powder
Chemical Name or Material L-Histidine Monohydrochloride
EINECSNumber 211-438-9
RTECSNumber MS3119000
TSCA Yes
Recommended Storage Ambient temperatures
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit