Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ L-Histidine Monohydrochloride Monohydrate, ≥98%, Thermo Scientific™

Click to view available options
Quantity:
100g
1kg
Packaging:
Plastic bottle
5934-29-2
C6H12ClN3O3
209.63
MFCD00151027
CMXXUDSWGMGYLZ-XRIGFGBMSA-N
l-histidine hydrochloride hydrate,l-histidine hydrochloride monohydrate,s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate,h-his-oh.hcl.h2o,l-histidine monohydrochloride monohydrate,l-histidine, monohydrochloride, monohydrate,l-histidin hydrate hydrochloride,histidine, monohydrochloride, monohydrate, l,h-his-ohhclh2o,l-histidine, hydrochloride, monohydrate
165377
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydrate;hydrochloride
C1=C(NC=N1)CC(C(=O)O)N.O.Cl
5934-29-2
C6H12ClN3O3
209.63
MFCD00151027
CMXXUDSWGMGYLZ-XRIGFGBMSA-N
l-histidine hydrochloride hydrate,l-histidine hydrochloride monohydrate,s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate,h-his-oh.hcl.h2o,l-histidine monohydrochloride monohydrate,l-histidine, monohydrochloride, monohydrate,l-histidin hydrate hydrochloride,histidine, monohydrochloride, monohydrate, l,h-his-ohhclh2o,l-histidine, hydrochloride, monohydrate
165377
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydrate;hydrochloride
C1=C(NC=N1)CC(C(=O)O)N.O.Cl
Linear Formula (C3H3N2)CH2CH(NH2)COOH.HCl.H2O
Packaging Plastic bottle
Formula Weight 209.63
Physical Form Crystalline Powder
Percent Purity ≥98%
Melting Point ~240°C (decomposition)
Quantity 1kg
Chemical Name or Material L-Histidine Monohydrochloride