Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-Histidine, Multi-Compendial, U.S.P., J.T.Baker™
Click to view available options
Quantity:
100g
1kg
Packaging:
Poly Bottle
Specifications
Packaging | Poly Bottle |
Formula Weight | 155.16 |
Grade | Multi-Compendial/USP |
Quantity | 1kg |
Specific Gravity | 1L = 1.44kg |
Chemical Name or Material | L-Histidine |