Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Histidine, Multi-Compendial, U.S.P., J.T.Baker™

Click to view available options
Quantity:
100g
1kg
Packaging:
Poly Bottle

Chemical Identifiers

71-00-1
C6H9N3O2
155.157
HNDVDQJCIGZPNO-YFKPBYRVSA-N
6274
CHEBI:15971
CAS 71-00-1
Molecular Formula C6H9N3O2
Packaging Poly Bottle
InChI Key HNDVDQJCIGZPNO-YFKPBYRVSA-N
SMILES C1=C(NC=N1)CC(C(=O)O)N
IUPAC Name (2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
Molecular Weight (g/mol) 155.157
PubChem CID 6274
ChEBI CHEBI:15971
Formula Weight 155.16
Quantity 100g
Specific Gravity 1L = 1.44kg
Chemical Name or Material L-Histidine
Grade Multi-Compendial/USP
Show More Show Less