Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Homophenylalanine, 98%

Manufacturer:  Alfa Aesar™ H2729514

Quantity 25g

 View more versions of this product

Catalog No. AAH2729514


  Check Availability
Add to cart

Specifications

Description

Applications
L-Homophenylalanine is used as a precursor in the pharmaceutical industry for the production of angiotensin-converting enzyme (ACE) and R-(-)-Homophenylalanin-ethylester. It is also involved in the synthesis of NEPA [(S,S)-N-(1-Ethoxycarbonyl-3-phenylpropyl)alanine], which is a key intermediate to prepare the ACE-inhibitors: enalapril, delapril, quinapril and ramipril. It acts as an anti-tumor reagent.

Solubility
Soluble in dilute aqueous acid.

Notes
Store in cool place. Incompatible with strong oxidizing agents, strong acids and bases.
Description & Specifications

Chemical Identifiers

943-73-7
179.22
(S)-2-Amino-4-phenylbutyric acid; H-Homophe-OH
C10H13NO2
MFCD00002619
L-Homophenylalanine

Specifications

943-73-7
179.22
(S)-2-Amino-4-phenylbutyric acid; H-Homophe-OH
25g
L-Homophenylalanine
C10H13NO2
MFCD00002619
>300°C
+45° (c=1 in 3N HCl)
98%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit