Learn More
L-Homophenylalanine, 98%, Thermo Scientific Chemicals
Description
L-Homophenylalanine is used as a precursor in the pharmaceutical industry for the production of angiotensin-converting enzyme (ACE) and R-(-)-Homophenylalanin-ethylester. It is also involved in the synthesis of NEPA [(S,S)-N-(1-Ethoxycarbonyl-3-phenylpropyl)alanine], which is a key intermediate to prepare the ACE-inhibitors: enalapril, delapril, quinapril and ramipril. It acts as an anti-tumor reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
L-Homophenylalanine is used as a precursor in the pharmaceutical industry for the production of angiotensin-converting enzyme (ACE) and R-(-)-Homophenylalanin-ethylester. It is also involved in the synthesis of NEPA [(S,S)-N-(1-Ethoxycarbonyl-3-phenylpropyl)alanine], which is a key intermediate to prepare the ACE-inhibitors: enalapril, delapril, quinapril and ramipril. It acts as an anti-tumor reagent.
Solubility
Soluble in dilute aqueous acid.
Notes
Store in cool place. Incompatible with strong oxidizing agents, strong acids and bases.
Specifications
Melting Point | >300°C |
Solubility Information | Soluble in dilute aqueous acid. |
IUPAC Name | (2S)-2-amino-4-phenylbutanoic acid |
Formula Weight | 179.22 |
Percent Purity | 98% |
Quantity | 1 g |
Optical Rotation | +45° (c=1 in 3N HCl) |
Chemical Name or Material | L-Homophenylalanine |
Safety and Handling
EINECSNumber | 213-403-3 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.