Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Homophenylalanine ethyl ester hydrochloride, 98%

Click to view available options
Quantity:
25g
100g
90891-21-7
C12H18ClNO2
243.731
MFCD00190691
PTFKZMFFSIYCOV-MERQFXBCSA-N
ethyl s-2-amino-4-phenylbutanoate hydrochloride,l-homophenylalanine ethyl ester hydrochloride,h-hophe-oet.hcl,ethyl l-homoalaninate hcl,s-ethyl 2-amino-4-phenylbutanoate hydrochloride,ethyl 2s-2-amino-4-phenylbutanoate hydrochloride,pubchem6054,h-hph-oet.hcl,h-hph-oet. hcl,homophenylalanine ethyl ester hydrochloride
2734606
ethyl (2S)-2-amino-4-phenylbutanoate;hydrochloride
CCOC(=O)C(CCC1=CC=CC=C1)N.Cl
90891-21-7
C12H18ClNO2
243.731
MFCD00190691
PTFKZMFFSIYCOV-MERQFXBCSA-N
ethyl s-2-amino-4-phenylbutanoate hydrochloride,l-homophenylalanine ethyl ester hydrochloride,h-hophe-oet.hcl,ethyl l-homoalaninate hcl,s-ethyl 2-amino-4-phenylbutanoate hydrochloride,ethyl 2s-2-amino-4-phenylbutanoate hydrochloride,pubchem6054,h-hph-oet.hcl,h-hph-oet. hcl,homophenylalanine ethyl ester hydrochloride
2734606
ethyl (2S)-2-amino-4-phenylbutanoate;hydrochloride
CCOC(=O)C(CCC1=CC=CC=C1)N.Cl
Beilstein 6448497
Formula Weight 243.73
Physical Form Powder
Percent Purity 98%
Color White
Melting Point 159°C to 163°C
Quantity 25g
Chemical Name or Material L-Homophenylalanine ethyl ester hydrochloride

storageNote1 : 2° to 8°C