Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Homoserine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
672-15-1
C4H9NO3
119.12
MFCD00063090
UKAUYVFTDYCKQA-VKHMYHEASA-N
l-homoserine,homoserine,s-2-amino-4-hydroxybutanoic acid,2s-2-amino-4-hydroxybutanoic acid,h-hoser-oh,s-2-amino-4-hydroxybutyric acid,homoserine van,2-amino-4-hydroxybutyric acid,s-homoserine,unii-6ka95x0ivo
12647
CHEBI:15699
(2S)-2-amino-4-hydroxybutanoic acid
C(CO)C(C(=O)O)N
672-15-1
C4H9NO3
119.12
MFCD00063090
UKAUYVFTDYCKQA-VKHMYHEASA-N
l-homoserine,homoserine,s-2-amino-4-hydroxybutanoic acid,2s-2-amino-4-hydroxybutanoic acid,h-hoser-oh,s-2-amino-4-hydroxybutyric acid,homoserine van,2-amino-4-hydroxybutyric acid,s-homoserine,unii-6ka95x0ivo
12647
CHEBI:15699
(2S)-2-amino-4-hydroxybutanoic acid
C(CO)C(C(=O)O)N
Assay Percent Range 97%
Infrared Spectrum Authentic
Beilstein 04, III, 1636
Merck Index 15, 4778
Packaging Glass bottle
Formula Weight 119.12
Specific Rotation -8° to -9° (22°C, 589nm) (c=2, H2O)
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Specific Rotation Condition − 8.50 (22.00°C c=2,H2O)
Color White to Yellow
Quantity 5g
Chemical Name or Material L-Homoserine
Show More Show Less