Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-alpha-Hydroxyisocaproic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
13748-90-8
C6H12O3
132.16
LVRFTAZAXQPQHI-YFKPBYRVSA-N
l-leucic acid,s-2-hydroxy-4-methylpentanoic acid,2s-2-hydroxy-4-methylpentanoic acid,l-2-hydroxy-4-methylvaleric acid,s-leucic acid,s-2-hydroxy-4-methylvaleric acid,l-2-hydroxyisocaproic acid,l-alpha-hydroxyisocaproic acid,s---2-hydroxyisocaproic acid,pentanoic acid, 2-hydroxy-4-methyl-, 2s
83697
CHEBI:44510
(2S)-2-hydroxy-4-methylpentanoic acid
CC(C)CC(C(=O)O)O
13748-90-8
C6H12O3
132.16
LVRFTAZAXQPQHI-YFKPBYRVSA-N
l-leucic acid,s-2-hydroxy-4-methylpentanoic acid,2s-2-hydroxy-4-methylpentanoic acid,l-2-hydroxy-4-methylvaleric acid,s-leucic acid,s-2-hydroxy-4-methylvaleric acid,l-2-hydroxyisocaproic acid,l-alpha-hydroxyisocaproic acid,s---2-hydroxyisocaproic acid,pentanoic acid, 2-hydroxy-4-methyl-, 2s
83697
CHEBI:44510
(2S)-2-hydroxy-4-methylpentanoic acid
CC(C)CC(C(=O)O)O
Packaging Glass Bottle
Quantity 1g
Melting Point 78°C to 80°C
Assay Percent Range 98%
Linear Formula (CH3)2CHCH2CH(OH)COOH
Beilstein 03,336
Formula Weight 132.16
Percent Purity 98%
Chemical Name or Material L-α-Hydroxyisocaproic acid

GHS Signal Word: Danger

EINECSNumber : 237-329-6