Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Isoleucinol, 97%, Thermo Scientific™

Click to view available options
Quantity:
1g
5g
24629-25-2
C6H15NO
117.192
MFCD00004731
VTQHAQXFSHDMHT-NTSWFWBYSA-N
l-isoleucinol, 2s,3s-2-amino-3-methylpentan-1-ol, s-+-isoleucinol, h-isoleucinol, l-+-isoleucinol, isoleucinol, 2s,3s-2-amino-3-methyl-pentan-1-ol, h-ile-ol, 1-pentanol, 2-amino-3-methyl-, 2s,3s
2724027
(2S,3S)-2-amino-3-methylpentan-1-ol
CCC(C)C(CO)N
24629-25-2
C6H15NO
117.192
MFCD00004731
VTQHAQXFSHDMHT-NTSWFWBYSA-N
l-isoleucinol, 2s,3s-2-amino-3-methylpentan-1-ol, s-+-isoleucinol, h-isoleucinol, l-+-isoleucinol, isoleucinol, 2s,3s-2-amino-3-methyl-pentan-1-ol, h-ile-ol, 1-pentanol, 2-amino-3-methyl-, 2s,3s
2724027
(2S,3S)-2-amino-3-methylpentan-1-ol
CCC(C)C(CO)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Refractive Index 1.4589
Melting Point 36°C to 39°C
Boiling Point 97°C
Quantity 5g
Flash Point 100°C (212°F)
Linear Formula C2H5CH(CH3)CH(NH2)CH2OH
Formula Weight 117.19
Percent Purity 97%
Chemical Name or Material L-Isoleucinol

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 246-371-4

TSCA : No

Recommended Storage : Ambient temperatures

RUO – Research Use Only