Alfa Aesar™ L-Isoleucyl diphenylborinate, 95%

Manufacturer: Alfa Aesar™ H55184MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH55184MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 124383-92-2
    Molecular Formula C18H22BNO2
    Formula Weight 295.19
    MDL Number MFCD22125121
    Synonym Diphenylborinic acid L-isoleucyl ester
    Chemical Name or Material L-Isoleucyl diphenylborinate

    Specifications

    Formula Weight 295.19
    Quantity 250mg
    Molecular Formula C18H22BNO2
    MDL Number MFCD22125121
    CAS 124383-92-2
    Synonym Diphenylborinic acid L-isoleucyl ester
    Chemical Name or Material L-Isoleucyl diphenylborinate
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit