Learn More
L-(+)-Lactic acid, ca 85-90% aq. soln., Thermo Scientific Chemicals
Description
It is most commonly used for fluid resuscitation after blood loss due to trauma, surgery, or burn injury. It is used in the agricultural, chemical, leather processing, pharmaceuticals, and cosmetics industries, used as an electroplating agent, food/feed additive, pH regulator, cleaning/washing agent, and tanning agent; used to flavor animal feeds; used as a solvent and intermediate in chemical production, as a pH regulator in fabric finishing, and in paints, coatings, soaps, and cleaning products; used to make large scale and fine chemicals, pulp-paper-paper products, food products, and plastic products, in mining, health services, agriculture-forestry-fishing, and building and construction work.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is most commonly used for fluid resuscitation after blood loss due to trauma, surgery, or burn injury. It is used in the agricultural, chemical, leather processing, pharmaceuticals, and cosmetics industries, used as an electroplating agent, food/feed additive, pH regulator, cleaning/washing agent, disinfectant, and tanning agent; used to flavor animal feeds; used as a solvent and intermediate in chemical production, as a pH regulator in fabric finishing, and in paints, coatings, soaps, and cleaning products; used to make large scale and fine chemicals, pulp-paper-paper products, food products, and plastic products, in mining, health services, agriculture-forestry-fishing, and building and construction work.
Solubility
Fully miscible in water. Soluble in water.
Notes
Store away from strong bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
pH | 2.8 |
Melting Point | 18°C |
Beilstein | 1720251 |
Merck Index | 145337 |
Solubility Information | Fully miscible in water. Soluble in water. |
Formula Weight | 90.08 |
Concentration or Composition (by Analyte or Components) | ≈85 to 90% aq. Soln. |
Density | 1.209 |
Quantity | 100 g |
Flash Point | >110°C (230°F) |
Show More |
Safety and Handling
Hazard Category | H315-H318 |
Hazard Statement | GHS H Statement H318-H315 Causes serious eye damage. Causes skin irritation. |
Precautionary Statement | P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362 |
EINECSNumber | 201-196-2 |
RTECSNumber | OD3100000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.