Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-(+)-Lactic acid lithium salt, Affymetrix/USB™

Click to view available options
Quantity:
100g
1kg
Packaging:
Plastic bottle
27848-80-2
C3H5LiO3
96.01
MFCD00065399
GKQWYZBANWAFMQ-DKWTVANSSA-M
lithium s-2-hydroxypropanoate,l-lactic acid lithium salt,lithium l-lactate,s-2-hydroxypropionic acid lithium salt,sarcolactic acid lithium salt,lithium 1+ l-+-lactate,lithiuml-lactate,lithotab l-lactate,lactic acid lithium salt,c3h5o3.li
23687877
lithium;(2S)-2-hydroxypropanoate
[Li+].CC(C(=O)[O-])O
27848-80-2
C3H5LiO3
96.01
MFCD00065399
GKQWYZBANWAFMQ-DKWTVANSSA-M
lithium s-2-hydroxypropanoate,l-lactic acid lithium salt,lithium l-lactate,s-2-hydroxypropionic acid lithium salt,sarcolactic acid lithium salt,lithium 1+ l-+-lactate,lithiuml-lactate,lithotab l-lactate,lactic acid lithium salt,c3h5o3.li
23687877
lithium;(2S)-2-hydroxypropanoate
[Li+].CC(C(=O)[O-])O
Packaging Plastic bottle
Quantity 1kg
Melting Point 253-255°
Formula Weight 96.01
Physical Form Crystalline powder
Chemical Name or Material L-(+)-Lactic acid lithium salt