Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Lactic Acid, Multi-Compendial, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
19L
Packaging:
Narrow Mouth Amber Glass Bottle
Poly Pail

Chemical Identifiers

79-33-4
C3H6O3
90.078
JVTAAEKCZFNVCJ-REOHCLBHSA-N
107689
CHEBI:422
SMILES CC(C(=O)O)O
IUPAC Name (2S)-2-hydroxypropanoic acid
Formula Weight 90.08
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.20kg
Quantity 4L
Flash Point 112.00°C
Chemical Name or Material L-Lactic Acid
Grade Multi-Compendial