Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Lactic Acid, Multi-Compendial, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
19L
Packaging:
Narrow Mouth Amber Glass Bottle
Poly Pail
79-33-4
C3H6O3
90.078
JVTAAEKCZFNVCJ-REOHCLBHSA-N
l-lactic acid,l-+-lactic acid,s-2-hydroxypropanoic acid,s-lactic acid,sarcolactic acid,s-2-hydroxypropionic acid,2s-2-hydroxypropanoic acid,+-lactic acid,espiritin,tisulac
107689
CHEBI:422
(2S)-2-hydroxypropanoic acid
CC(C(=O)O)O
79-33-4
C3H6O3
90.078
JVTAAEKCZFNVCJ-REOHCLBHSA-N
l-lactic acid,l-+-lactic acid,s-2-hydroxypropanoic acid,s-lactic acid,sarcolactic acid,s-2-hydroxypropionic acid,2s-2-hydroxypropanoic acid,+-lactic acid,espiritin,tisulac
107689
CHEBI:422
(2S)-2-hydroxypropanoic acid
CC(C(=O)O)O
Formula Weight 90.08
Grade Multi-Compendial
Packaging Poly Pail
Specific Gravity 1L = 1.20kg
Quantity 19L
Flash Point 112.00°C
Chemical Name or Material L-Lactic Acid