Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Lactic acid sodium salt, 99%, extra pure, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
867-56-1
C3H5NaO3
112.06
sodium l-lactate,sodium-l-lactate,sodium lactate, l,sodium s-2-hydroxypropanoate,unii-p2y1c6m9ps,sodium s-lactate,sodium l-+-lactate,p2y1c6m9ps,propanoic acid, 2-hydroxy-, monosodium salt, 2s,l-+-lactic acid sodium salt
867-56-1
C3H5NaO3
112.06
sodium l-lactate,sodium-l-lactate,sodium lactate, l,sodium s-2-hydroxypropanoate,unii-p2y1c6m9ps,sodium s-lactate,sodium l-+-lactate,p2y1c6m9ps,propanoic acid, 2-hydroxy-, monosodium salt, 2s,l-+-lactic acid sodium salt
Assay Percent Range 99%
Linear Formula CH3CH(OH)CO2Na
Formula Weight 112.06
Physical Form Powder
Percent Purity 99%
Grade Extra Pure
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Rotation −12° to −13° (20°C, 589nm) (C=1 in H2O)
Water 1% Max.
Color White to Yellow
Melting Point 161.6°C to 165°C
Quantity 10g
Chemical Name or Material L-Lactic acid sodium salt, Extra Pure
Show More Show Less

EINECSNumber : 212-762-3