Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Leucinamide hydrochloride, 99%

Click to view available options
Quantity:
1g
5g
10466-61-2
C6H15ClN2O
166.649
MFCD00013012
VSPSRRBIXFUMOU-JEDNCBNOSA-N
l-leucinamide hydrochloride,h-leu-nh2.hcl,s-2-amino-4-methylpentanamide hydrochloride,l-leucinamide hcl,2s-2-amino-4-methylpentanamide hydrochloride,pentanamide, 2-amino-4-methyl-, monohydrochloride, 2s,c6h14n2o.hcl,l-leucine amide hydrochloride,leu-nh2.hcl,h-d-leu-nh hcl
2733199
(2S)-2-amino-4-methylpentanamide;hydrochloride
CC(C)CC(C(=O)N)N.Cl
10466-61-2
C6H15ClN2O
166.649
MFCD00013012
VSPSRRBIXFUMOU-JEDNCBNOSA-N
l-leucinamide hydrochloride,h-leu-nh2.hcl,s-2-amino-4-methylpentanamide hydrochloride,l-leucinamide hcl,2s-2-amino-4-methylpentanamide hydrochloride,pentanamide, 2-amino-4-methyl-, monohydrochloride, 2s,c6h14n2o.hcl,l-leucine amide hydrochloride,leu-nh2.hcl,h-d-leu-nh hcl
2733199
(2S)-2-amino-4-methylpentanamide;hydrochloride
CC(C)CC(C(=O)N)N.Cl
Quantity 1g
Optical Rotation +9.5° (c=2 in Water)
Melting Point ~240°C (decomposition)
Beilstein 4237021
Formula Weight 166.65
Percent Purity 99%
Chemical Name or Material L-Leucinamide hydrochloride