Alfa Aesar™ L-Leucinamide hydrochloride, 99%

Manufacturer: Alfa Aesar™ B2291406

Quantity 5g

 View more versions of this product

Catalog No. AAB2291406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10466-61-2
    Molecular Formula C6H14N2O·HCl
    Formula Weight 166.65
    MDL Number MFCD00013012
    Beilstein 4237021
    Synonym (R)-2-Amino-4-methylvaleramide; H-Leu-NH2
    Chemical Name or Material L-Leucinamide hydrochloride

    Specifications

    Formula Weight 166.65
    Molecular Formula C6H14N2O·HCl
    MDL Number MFCD00013012
    CAS 10466-61-2
    Beilstein 4237021
    Synonym (R)-2-Amino-4-methylvaleramide; H-Leu-NH2
    Melting Point ≈240°C (dec.)
    Quantity 5g
    Optical Rotation +9.5° (c=2 in Water)
    Chemical Name or Material L-Leucinamide hydrochloride
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit