Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Leucine benzyl ester p-toluenesulfonate, 98%

Click to view available options
Quantity:
1g
5g
25g
1738-77-8
C20H27NO5S
393.498
MFCD00066117
QTQGHKVYLQBJLO-YDALLXLXSA-N
l-leucine benzyl ester p-toluenesulfonate salt,l-leucine benzyl ester p-toluenesulfonate,h-leu-obzl.tosoh,l-leucine benzyl ester tosylate,o-benzyl-l-leucine toluene-p-sulphonate,s-benzyl 2-amino-4-methylpentanoate 4-methylbenzenesulfonate,benzyl 2s-2-amino-4-methylpentanoate; para-toluene sulfonate,leu-obzl tosoh,h-leu-obzl.tos,h-leu-obzl tsoh
6451312
benzyl (2S)-2-amino-4-methylpentanoate;4-methylbenzenesulfonic acid
CC1=CC=C(C=C1)S(=O)(=O)O.CC(C)CC(C(=O)OCC1=CC=CC=C1)N
1738-77-8
C20H27NO5S
393.498
MFCD00066117
QTQGHKVYLQBJLO-YDALLXLXSA-N
l-leucine benzyl ester p-toluenesulfonate salt,l-leucine benzyl ester p-toluenesulfonate,h-leu-obzl.tosoh,l-leucine benzyl ester tosylate,o-benzyl-l-leucine toluene-p-sulphonate,s-benzyl 2-amino-4-methylpentanoate 4-methylbenzenesulfonate,benzyl 2s-2-amino-4-methylpentanoate; para-toluene sulfonate,leu-obzl tosoh,h-leu-obzl.tos,h-leu-obzl tsoh
6451312
benzyl (2S)-2-amino-4-methylpentanoate;4-methylbenzenesulfonic acid
CC1=CC=C(C=C1)S(=O)(=O)O.CC(C)CC(C(=O)OCC1=CC=CC=C1)N
Beilstein 3566840
Formula Weight 393.5
Percent Purity 98%
Melting Point 156°C to 160°C
Quantity 1g
Chemical Name or Material L-Leucine benzyl ester p-toluenesulfonate

EINECSNumber : 217-095-1

TSCA : No

Recommended Storage : Ambient temperatures