Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Leucine ethyl ester hydrochloride, 99%, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2743-40-0
C8H17NO2·ClH
195.69
NOUDPBCEONUCOV-FJXQXJEOSA-N
l-leucine ethyl ester hydrochloride,ethyl l-leucinate hydrochloride,h-leu-oet.hcl,s-ethyl 2-amino-4-methylpentanoate hydrochloride,l-leucine ethyl ester hcl,h-leu-oet inverted exclamation mark currencyhcl,leucine, ethyl ester hydrochloride,h-leu-oet,leucine ethyl ester hydrochloride,leu-oet hcl
11658447
ethyl (2S)-2-amino-4-methylpentanoate;hydrochloride
CCOC(=O)C(CC(C)C)N.Cl
2743-40-0
C8H17NO2·ClH
195.69
NOUDPBCEONUCOV-FJXQXJEOSA-N
l-leucine ethyl ester hydrochloride,ethyl l-leucinate hydrochloride,h-leu-oet.hcl,s-ethyl 2-amino-4-methylpentanoate hydrochloride,l-leucine ethyl ester hcl,h-leu-oet inverted exclamation mark currencyhcl,leucine, ethyl ester hydrochloride,h-leu-oet,leucine ethyl ester hydrochloride,leu-oet hcl
11658447
ethyl (2S)-2-amino-4-methylpentanoate;hydrochloride
CCOC(=O)C(CC(C)C)N.Cl
Melting Point 135°C to 137°C
Specific Rotation 15
Packaging Glass bottle
Quantity 5g
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Formula Weight 195.69
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material L-Leucine ethyl ester hydrochloride
Show More Show Less

EINECSNumber : 220-375-6