Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Lysinamide dihydrochloride, 95%

Click to view available options
Quantity:
250mg
1g
51127-08-3
C6H17Cl2N3O
218.122
MFCD00058287
AIYVJLPYZQDCKV-XRIGFGBMSA-N
s-2,6-diaminohexanamide dihydrochloride,h-lys-nh2 2hcl,l-lysinamide dihydrochloride,h-lys-nh 2 hcl,lysine amide dihydrochloride,c6h15n3o.2hcl,h-lys-nh2 . 2 hcl
20760365
(2S)-2,6-diaminohexanamide;dihydrochloride
C(CCN)CC(C(=O)N)N.Cl.Cl
51127-08-3
C6H17Cl2N3O
218.122
MFCD00058287
AIYVJLPYZQDCKV-XRIGFGBMSA-N
s-2,6-diaminohexanamide dihydrochloride,h-lys-nh2 2hcl,l-lysinamide dihydrochloride,h-lys-nh 2 hcl,lysine amide dihydrochloride,c6h15n3o.2hcl,h-lys-nh2 . 2 hcl
20760365
(2S)-2,6-diaminohexanamide;dihydrochloride
C(CCN)CC(C(=O)N)N.Cl.Cl
Formula Weight 218.12
Physical Form Solid
Percent Purity 95%
Color White
Quantity 1g
Chemical Name or Material L-Lysinamide dihydrochloride

storageNote1 : 2° to 8°C