Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Lysine acetate, 97%, Acros Organics

Click to view available options
Quantity:
25g
100g
500g
57282-49-2
C8H18N2O4
206.242
RRNJROHIFSLGRA-JEDNCBNOSA-N
l-lysine acetate,l-lysine acetate salt,lysine acetate,l-lysine monoacetate,unii-ttl6g7liwz,lysine acetate salt,lysine, monoacetate,lysine acetate usp,l-lysine, acetate,ttl6g7liwz
104152
acetic acid;(2S)-2,6-diaminohexanoic acid
CC(=O)O.C(CCN)CC(C(=O)O)N
57282-49-2
C8H18N2O4
206.242
RRNJROHIFSLGRA-JEDNCBNOSA-N
l-lysine acetate,l-lysine acetate salt,lysine acetate,l-lysine monoacetate,unii-ttl6g7liwz,lysine acetate salt,lysine, monoacetate,lysine acetate usp,l-lysine, acetate,ttl6g7liwz
104152
acetic acid;(2S)-2,6-diaminohexanoic acid
CC(=O)O.C(CCN)CC(C(=O)O)N
Assay Percent Range 97%
Formula Weight 206.24
Percent Purity 97%
Quantity 500g
Chemical Name or Material L-Lysine acetate