Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ L-Lysine hydrochloride, ≥98.5% (Dry Basis), Ultrapure, Thermo Scientific™

Click to view available options
Quantity:
1kg
5kg
10kg
Packaging:
Fiber drum
Plastic bottle
Poly Pail
657-27-2
C6H15ClN2O2
182.648
MFCD00064564
BVHLGVCQOALMSV-JEDNCBNOSA-N
l-lysine hydrochloride,l-lysine monohydrochloride,lysine hydrochloride,h-lys-oh.hcl,l-lysine, monohydrochloride,lyamine,darvyl,lysion,s-2,6-diaminohexanoic acid hydrochloride,enisyl
69568
CHEBI:53633
(2S)-2,6-diaminohexanoic acid;hydrochloride
C(CCN)CC(C(=O)O)N.Cl
657-27-2
C6H15ClN2O2
182.648
MFCD00064564
BVHLGVCQOALMSV-JEDNCBNOSA-N
l-lysine hydrochloride,l-lysine monohydrochloride,lysine hydrochloride,h-lys-oh.hcl,l-lysine, monohydrochloride,lyamine,darvyl,lysion,s-2,6-diaminohexanoic acid hydrochloride,enisyl
69568
CHEBI:53633
(2S)-2,6-diaminohexanoic acid;hydrochloride
C(CCN)CC(C(=O)O)N.Cl
Linear Formula NH2(CH2)4CH(NH2)COOH.HCl
Packaging Plastic bottle
Formula Weight 182.65
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Melting Point ~260°C (decomposition)
Quantity 1kg
Chemical Name or Material L-Lysine hydrochloride
Assay Dry basis