Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L(-)-Malic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Plastic bottle
97-67-6
C4H6O5
134.087
MFCD00064213
BJEPYKJPYRNKOW-REOHCLBHSA-N
l-malic acid,l---malic acid,s-2-hydroxysuccinic acid,2s-2-hydroxybutanedioic acid,l--malic acid,apple acid,--malic acid,l-apple acid,s-malic acid,s-2-hydroxybutanedioic acid
222656
CHEBI:30797
(2S)-2-hydroxybutanedioic acid
C(C(C(=O)O)O)C(=O)O
97-67-6
C4H6O5
134.087
MFCD00064213
BJEPYKJPYRNKOW-REOHCLBHSA-N
l-malic acid,l---malic acid,s-2-hydroxysuccinic acid,2s-2-hydroxybutanedioic acid,l--malic acid,apple acid,--malic acid,l-apple acid,s-malic acid,s-2-hydroxybutanedioic acid
222656
CHEBI:30797
(2S)-2-hydroxybutanedioic acid
C(C(C(=O)O)O)C(=O)O
Additional Information fumaric acid: < 0.2%
Arsenic (As) 2ppm max.
Assay Percent Range 99%
Linear Formula HO2CCH2CH(OH)CO2H
Chlorine (Cl) 100ppm max.
Beilstein 03, 419
Merck Index 15, 5771
Formula Weight 134.09
Physical Form Crystalline Powder or Granules
Percent Purity ≥98.5%
Heavy Metals (as Pb) 20ppm max.
Infrared Spectrum Authentic
Packaging Plastic bottle
Specific Rotation -1.5° to -3° (20°C, 589nm) (c=8.5, water)
Sulfate 300ppm max.
Sulfated Ash 0.2% max.
Specific Rotation Condition − 2.00 (20.00°C c=8.5,H2O)
Melting Point 99°C to 107°C
Color White
Quantity 25g
Chemical Name or Material L(-)-Malic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger