Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L(-)-Menthol, 99.5%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
Plastic Bottle
2216-51-5
C10H20O
156.27
MFCD00062979
NOOLISFMXDJSKH-KXUCPTDWSA-N
l-menthol,--menthol,levomenthol,menthomenthol,l---menthol,hexahydrothymol,menthacamphor,peppermint camphor,1r,2s,5r-2-isopropyl-5-methylcyclohexanol,u.s.p. menthol
16666
CHEBI:15409
(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-ol
CC1CCC(C(C1)O)C(C)C
2216-51-5
C10H20O
156.27
MFCD00062979
NOOLISFMXDJSKH-KXUCPTDWSA-N
l-menthol,--menthol,levomenthol,menthomenthol,l---menthol,hexahydrothymol,menthacamphor,peppermint camphor,1r,2s,5r-2-isopropyl-5-methylcyclohexanol,u.s.p. menthol
16666
CHEBI:15409
(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-ol
CC1CCC(C(C1)O)C(C)C
Specific Rotation Condition −51.00 (20.00°C 589nm,c=10,EtOH)
Refractive Index 1.46
Specific Rotation − 51.00
Infrared Spectrum Authentic
Packaging Plastic Bottle
Melting Point 41.0°C to 43.0°C
Flash Point 94°C
Quantity 2.5kg
Specific Gravity 0.89
Boiling Point 212.0°C
Density 0.8900g/mL
Color Colorless to White
Assay Percent Range 99.5%
Beilstein 06,28
Fieser 13,172; 16,203
Merck Index 15,5905
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in acetic acid,soluble in alcohol,chloroform,ether,,petroleum ether
Formula Weight 156.27
Physical Form Crystals or Crystalline Needles
Percent Purity 99.5%
Chemical Name or Material L(-)-Menthol, 99.7%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 218-690-9

RTECSNumber : OT0700000

TSCA : TSCA