Alfa Aesar™ L-Methionine ethyl ester hydrochloride, 99%

Manufacturer: Alfa Aesar™ B25055-14

Quantity 25g

 View more versions of this product

Catalog No. AAB2505514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2899-36-7
    Molecular Formula C7H15NO2S·HCl
    Formula Weight 213.73g/mol
    MDL Number MFCD00012508
    Synonym H-Met-OEt·HCl
    Chemical Name or Material L-Methionine ethyl ester hydrochloride

    Specifications

    Melting Point 90° to 92°C
    Quantity 25g
    CAS 2899-36-7
    Molecular Formula C7H15NO2S·HCl
    Formula Weight 213.73g/mol
    MDL Number MFCD00012508
    Synonym H-Met-OEt·HCl
    Chemical Name or Material L-Methionine ethyl ester hydrochloride
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit