Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Methionine ethyl ester hydrochloride, 99%

Manufacturer:  Alfa Aesar™ B2505522

Quantity 100g

 View more versions of this product

Catalog No. AAB2505522


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

2899-36-7
213.73
H-Met-OEt·HCl
C7H15NO2S·HCl
MFCD00012508
L-Methionine ethyl ester hydrochloride

Specifications

2899-36-7
213.73
H-Met-OEt·HCl
100g
99%
C7H15NO2S·HCl
MFCD00012508
90° to 92°C
L-Methionine ethyl ester hydrochloride
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit