Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Methionine ethyl ester hydrochloride, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
2899-36-7
C7H15NO2S·HCl
213.73
KPWCQEUBJAIORR-RGMNGODLSA-N
l-methionine ethyl ester hydrochloride,ethyl l-methionate hydrochloride,h-met-oet.hcl,ethyll-methionatehydrochloride,l-methionine, ethyl ester, hydrochloride,h-met-oet hcl,ethyl l-methionate hcl,ethyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride,h-met-oet?cl,pubchem14924
3083785
ethyl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride
CCOC(=O)C(CCSC)N.Cl
2899-36-7
C7H15NO2S·HCl
213.73
KPWCQEUBJAIORR-RGMNGODLSA-N
l-methionine ethyl ester hydrochloride,ethyl l-methionate hydrochloride,h-met-oet.hcl,ethyll-methionatehydrochloride,l-methionine, ethyl ester, hydrochloride,h-met-oet hcl,ethyl l-methionate hcl,ethyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride,h-met-oet?cl,pubchem14924
3083785
ethyl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride
CCOC(=O)C(CCSC)N.Cl
Specific Rotation Condition + 21.00 (25.00°C c=2,EtOH)
Packaging Glass bottle
Quantity 10g
Specific Rotation + 21.00
Melting Point 82.0°C to 92.0°C
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula CH3SCH2CH2CH(NH2)CO2C2H5·HCl
Formula Weight 213.73
Percent Purity 99%
Chemical Name or Material L-Methionine ethyl ester hydrochloride, 99%

GHS Signal Word: Warning

EINECSNumber : 220-787-6