Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Methionine, Multi-Compendial, U.S.P., J.T.Baker™

Click to view available options
Quantity:
100g
1kg
Packaging:
Poly Bottle
63-68-3
C5H11NO2S
149.208
FFEARJCKVFRZRR-BYPYZUCNSA-N
l-methionine,methionine,h-met-oh,s-2-amino-4-methylthio butanoic acid,cymethion,l---methionine,liquimeth,s-methionine,l-methioninum,methilanin
6137
CHEBI:16643
(2S)-2-amino-4-methylsulfanylbutanoic acid
CSCCC(C(=O)O)N
63-68-3
C5H11NO2S
149.208
FFEARJCKVFRZRR-BYPYZUCNSA-N
l-methionine,methionine,h-met-oh,s-2-amino-4-methylthio butanoic acid,cymethion,l---methionine,liquimeth,s-methionine,l-methioninum,methilanin
6137
CHEBI:16643
(2S)-2-amino-4-methylsulfanylbutanoic acid
CSCCC(C(=O)O)N
Packaging Poly Bottle
Formula Weight 149.21
Grade Multi-Compendial/USP
Quantity 1kg
Chemical Name or Material L-Methionine